Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRCCHICKLP GRVMGIRVLR LSLVVILVLL LVAGALTALL PSVKEDKMLM LRREIKSQGK STMDSFTLIM QTYNRTDLLL KLLNHYQAVP NLHKVIVVWN NIGEKAPDEL WNSLGPHPIP VIFKQQTANR MRNRLQVFPE LETNAVLMVD DDTLISTPDL VFAFSVWQQF PDQIVGFVPR KHVSTSSGIY SYGSFEMQAP GSGNGDQYSM VLIGASFFNS KYLELFQRQP AAVHALIDDT QNCDDIAMNF IIAKHIGKTS GIFVKPVNMD NLEKETNSGY SGMWHRAEHA LQRSYCINKL VNIYDSMPLR YSNIMISQFG FPYANYKRKI
EXTL2 is a glycosyltransferase required for the biosynthesis of heparan-sulfate and responsible for the alternating addition of beta-1-4-linked glucuronic acid (GlcA) and alpha-1-4-linked N-acetylglucosamine (GlcNAc) units to nascent heparan sulfate chains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-GalNAcT EXTL2; exostoses (multiple)-like 2; Exostosin-like 2; exostosin-like glycosyltransferase 2; EXT-related protein 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; processed exostosin-like 2
Gene Aliases: EXTL2; EXTR2
UniProt ID: (Human) Q9UBQ6
Entrez Gene ID: (Human) 2135
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support