Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: U87-MG, MCF-7.
Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (95%), Xenopus laevis (96%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EXTL3 is probable glycosyltransferase.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: exostoses (multiple)-like 3; exostosin tumor-like 3; Exostosin-like 3; exostosin-like glycosyltransferase 3; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Hereditary multiple exostoses gene isolog; Multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; reg receptor
Gene Aliases: BOTV; EXTL1L; EXTL3; EXTR1; KIAA0519; REGR; RPR
UniProt ID: (Human) O43909
Entrez Gene ID: (Human) 2137
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support