Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total EXTL3 protein.
EXTL3 is probable glycosyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: exostoses (multiple)-like 3; exostoses-like 3; exostosin; exostosin tumor-like 3; Exostosin-like 3; exostosin-like glycosyltransferase 3; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Hereditary multiple exostoses gene isolog; Multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; reg receptor
Gene Aliases: 2900009G18Rik; AU040105; BOTV; Ext1l; EXTL1L; EXTL3; EXTR1; KIAA0519; mKIAA0519; REGR; RPR
UniProt ID: (Human) O43909
Entrez Gene ID: (Human) 2137, (Mouse) 54616
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support