Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Eyes absent homolog 3 (EYA3) is a member of the eyes absent (EYA) family of proteins. This protein may act as a transcriptional activator and have a role during development. A similar protein in mice can act as a transcriptional activator. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eyes absent 3; eyes absent 3 homolog; Eyes absent homolog 3
Gene Aliases: AI844637; EYA3
UniProt ID: (Human) Q99504, (Mouse) P97480
Entrez Gene ID: (Human) 2140, (Rat) 313027, (Mouse) 14050
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support