Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: LTEDLKTIK QTHSQELCKL MNLWTERFCA LEEKCENIQK PLSSVQENIQ QKSKDIVNKM TFHSQKFCAD SDGFSQELRN FNQEGTKLVE ESVKHSDKLN GNLEKISQET EQRCESLNTR TVYFSEQWVS SLNEREQELH NLLEVVSQCC EASSSDITEK SDGRKAAHEK QHNIFLDQMT IDEDKLIAQN LELNETIKIG LTKLNCFLEQ DLKLDIPTGT TPQRKSYLYP STLVRTEPRE HLLDQLKRKQ PELLMMLNCS ENNKEETIPD VDVEEAVLGQ YTEEPLSQEP SVDAGVDCSS IGGVPFFQHK KSHGKDKENR GINTLERSKV EETTEHLVTK SRLPLRA (707-1052 aa encoded by BC136474)
This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kines; Kinesin-like protein 1; Kinesin-like protein KIF11; Kinesin-like spindle protein HKSP; Kinesin-related motor protein Eg5; Thyroid receptor-interacting protein 5; TR-interacting protein 5; TRIP-5
Gene Aliases: EG5; HKSP; KIF11; KNSL1; MCLMR; TRIP5
UniProt ID: (Human) P52732
Entrez Gene ID: (Human) 3832
Molecular Function: microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support