Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: DGSAPDSPFT SPPLREEIMA NNFSLESHNI SLTEHSSMPV EKNITLERPS NVNLTCQFTT SGDLNAVNVT WKKDGEQLEN NYLVSATGST LYTQYRFTII NSKQMGSYSC FFREEKEQRG TFNFKVPELH GKNKPLISYV GDSTVLTCKC QNCFPLNWTW YSSNGSVKVP VGVQMNKYVI NGTYANETKL KITQLLEEDG ESYWCRALFQ LGESEEHIEL VVLSYLVP; Positive Samples: Jurkat, MCF-7, THP-1; Cellular Location: Cell junction, Cell membrane, Single-pass type I membrane protein, synapse
Embigin is a 327 amino acid regulatory protein belonging to the immunoglobulin superfamily class of CAMs, and has two Ig-like (immunoglobulin-like) V- type domains. In situ hybridization analysis shows that Embigin has specific carbohydrate epitopes and plays an important role in early postimplantation embryogenesis. A highly glycosylated transmembrane glycoprotein, Embigin acts as a regulator of cell/ECM interactions during development and homeostasis of normal adult tissues. Transfection studies suggest its role in promotion of integrin-mediated cell-substratum adhesion. Widely expressed in most of the tissues and embryonal carcinoma cells, prostate and mammary cancer cell lines, it may also help in the classification of human carcinomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Embigin; embigin homolog
Gene Aliases: EMB; GP70
UniProt ID: (Human) Q6PCB8
Entrez Gene ID: (Human) 133418
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support