Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the endothelin family. Endothelins are endothelium-derived vasoactive peptides involved in a variety of biological functions. The active form of this protein is a 21 amino acid peptide processed from the precursor protein. The active peptide is a ligand for endothelin receptor type B (EDNRB). The interaction of this endothelin with EDNRB is essential for development of neural crest-derived cell lineages, such as melanocytes and enteric neurons. Mutations in this gene and EDNRB have been associated with Hirschsprung disease (HSCR) and Waardenburg syndrome (WS), which are congenital disorders involving neural crest-derived cells. Four alternatively spliced transcript variants encoding three distinct isoforms have been observed.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endothelin-3; ET 3; ET-3; MGC15067; MGC61498; PPET3; Preproendothelin 3; Preproendothelin-3; RP4-614C15.1
Gene Aliases: EDN3; ET-3; ET3; HSCR4; PPET3; WS4B
UniProt ID: (Human) P14138
Entrez Gene ID: (Human) 1908
Molecular Function: peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support