Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: EDNRB.
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AU022549; endothelin B receptor; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; endothelin-B receptor; ET-B; Et-b receptor; Et-b-r; ET-BR; ET-RB; Etb receptor; Etb-svr; piebald; RP11-318G21.1
Gene Aliases: ABCDS; Ednra; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETR-b; ETRB; HSCR; HSCR2; Sox10m1; WS4A
UniProt ID: (Human) P24530, (Rat) P21451, (Mouse) P48302
Entrez Gene ID: (Human) 1910, (Rat) 50672, (Mouse) 13618
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support