Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CLSRQELFP FGPGQGDLEL EDGDDFVSPA LELRGALRFY DRSDIDAVYV TTNGIIATSE PPAKESHPGL FPPTFGAVAP FLADLDTTDG LGKVYYREDL SPSITQRAAE CVHRGFPEIS FQPSSAVVVT WESVAPYQGP SRDPDQKGKR NTFQAVLASS DSSSYAIFLY PEDGLQFHTT FSKKENNQVP AVVAFSQGSV GFLWKSNGAY NIFANDRESI ENLAKSSNSG QQGVWVFEIG SPATTNGVVP ADVILGTEDG AEYDDEDEDY DLATTRLGLE DVGTTPFSYK ALRRGGADTY SVPSVLSPRR AATERPLGPP TERTRSFQLA VETFHQQHPQ VIDVDEVEET GVVFSYNTDS RQTCANNRHQ CSVH (28-400 aa encoded by BC045606)
This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enactin; Entactin; entactin 1; NID 1; NID-1; Nidogen (enactin); nidogen (entactin); Nidogen precursor (Entactin); Nidogen-1; Nidogen-1;
Gene Aliases: A630025O17; Ent; entactin; entactin-1; NID; NID1; nidogen-1
UniProt ID: (Human) P14543, (Mouse) P10493
Entrez Gene ID: (Human) 4811, (Rat) 25494, (Mouse) 18073
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support