Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml.
Enterokinase is an intestinal enzyme responsible for initiating activation of pancreatic proteolytic proenzymes. Mouse Anti-Enterokinase Monoclonal Antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified recombinant EK protein (bovine EK, aa848-1035, light chain, 93% homologous with human EK).Mouse Anti-Enterokinase Monoclonal Antibody recognizes EK in Western blots and ELISAs.
This gene encodes an enzyme that converts the pancreatic proenzyme trypsinogen to trypsin, which activates other proenzymes including chymotrypsinogen and procarboxypeptidases. The precursor protein is cleaved into two chains that form a heterodimer linked by a disulfide bond. This protein is a member of the trypsin family of peptidases. Mutations in this gene cause enterokinase deficiency, a malabsorption disorder characterized by diarrhea and failure to thrive. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enterokinase; Enteropeptidase; enteropeptidase, light chain (L chain); protease, serine, 7 (enterokinase); Serine protease 7; Transmembrane protease serine 15; transmembrane protease, serine 15
Gene Aliases: ENTK; PRSS7; TMPRSS15
UniProt ID: (Bovine) P98072, (Pig) P98074
Entrez Gene ID: (Bovine) 282009, (Pig) 397152
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support