Search
Search
Peptide sequence: AATPTPTPPT RKTPESFLGP NAALVDLDSL VSRPGPTPPG AKASNPFLPG
Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%
This gene encodes a member of the epsin protein family. The encoded protein binds clathrin and is involved in the endocytosis of clathrin-coated vesicles. Loss of function of this gene is associated with reduced tumor growth and progression in certain cancer types.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EH domain-binding mitotic phosphoprotein; EPS-15-interacting protein 1; Epsin-1
Gene Aliases: EPN1
UniProt ID: (Human) Q9Y6I3
Entrez Gene ID: (Human) 29924
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support