Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the epsin protein family. The encoded protein binds clathrin and is involved in the endocytosis of clathrin-coated vesicles. Loss of function of this gene is associated with reduced tumor growth and progression in certain cancer types.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EH domain-binding mitotic phosphoprotein; EPS-15-interacting protein 1; Epsin-1; Ibp1; Intersectin-EH-binding protein 1
Gene Aliases: AA269831; EPN1; Ibp1
UniProt ID: (Human) Q9Y6I3, (Mouse) Q80VP1
Entrez Gene ID: (Human) 29924, (Mouse) 13854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support