Invitrogen
FIGURE: 1 / 4
MA1-23214 detects Estrogen Receptor Alpha in Human samples.
MA1-23214 has been successfully used in Immunoprecipitation and Western Blot procedures. Positive control of MCF7 suggested.
MA1-23214 immunogen corresponds to The region encoding amino acids 1-190 expressed in E. coli.
Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze/thaw cycles.
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.