Product References
The absence of oestrogen receptor beta disturbs collagen I type deposition during Achilles tendon healing by regulating the IRF5-CCL3 axis.
Journal of cellular and molecular medicine
Bian X,Liu T,Yang M,Gu C,He G,Zhou M,Tang H,Lu K,Lai F,Wang F,Yang Q,Gustafsson JÅ,Fan X,Tang K
Published figure using Estrogen Receptor beta polyclonal antibody (Product # PA1-313) in Immunocytochemistry
Tue Sep 01 00:00:00 EDT 2020
The absence of oestrogen receptor beta disturbs collagen I type deposition during Achilles tendon healing by regulating the IRF5-CCL3 axis.
Journal of cellular and molecular medicine
Bian X,Liu T,Yang M,Gu C,He G,Zhou M,Tang H,Lu K,Lai F,Wang F,Yang Q,Gustafsson JÅ,Fan X,Tang K
Published figure using Estrogen Receptor beta polyclonal antibody (Product # PA1-313) in Immunocytochemistry
Tue Sep 01 00:00:00 EDT 2020
The absence of oestrogen receptor beta disturbs collagen I type deposition during Achilles tendon healing by regulating the IRF5-CCL3 axis.
Journal of cellular and molecular medicine
Bian X,Liu T,Yang M,Gu C,He G,Zhou M,Tang H,Lu K,Lai F,Wang F,Yang Q,Gustafsson JÅ,Fan X,Tang K
Published figure using Estrogen Receptor beta polyclonal antibody (Product # PA1-313) in Immunocytochemistry
Tue Sep 01 00:00:00 EDT 2020
Insights into the Role of Estrogen Receptor β in Triple-Negative Breast Cancer.
Cancers
Sellitto A,D'Agostino Y,Alexandrova E,Lamberti J,Pecoraro G,Memoli D,Rocco D,Coviello E,Giurato G,Nassa G,Tarallo R,Weisz A,Rizzo F
PA1-313 was used in Western Blot, Immunohistochemistry to comprehensively review the current state of knowledge concerning the role of estrogen receptor β in triple-negative breast cancer biology.
Fri Jun 05 00:00:00 EDT 2020
Insights into the Role of Estrogen Receptor β in Triple-Negative Breast Cancer.
Cancers
Sellitto A,D'Agostino Y,Alexandrova E,Lamberti J,Pecoraro G,Memoli D,Rocco D,Coviello E,Giurato G,Nassa G,Tarallo R,Weisz A,Rizzo F
PA1-313 was used in Western Blot, Immunohistochemistry to comprehensively review the current state of knowledge concerning the role of estrogen receptor β in triple-negative breast cancer biology.
Fri Jun 05 00:00:00 EDT 2020
Interaction Proteomics Identifies ERbeta Association with Chromatin Repressive Complexes to Inhibit Cholesterol Biosynthesis and Exert An Oncosuppressive Role in Triple-negative Breast Cancer.
Molecular & cellular proteomics : MCP
Alexandrova E,Giurato G,Saggese P,Pecoraro G,Lamberti J,Ravo M,Rizzo F,Rocco D,Tarallo R,Nyman TA,Collina F,Cantile M,Di Bonito M,Botti G,Nassa G,Weisz A
PA1-313 was used in Immunoprecipitation to explore the potential of targeting Estrogen Receptor beta (ERβ) and its role in triple-negative breast cancer cells.
Sat Feb 01 00:00:00 EST 2020
Absence of estrogen receptor beta leads to abnormal adipogenesis during early tendon healing by an up-regulation of PPARγ signalling.
Journal of cellular and molecular medicine
Bian X,Liu T,Zhou M,He G,Ma Y,Shi Y,Wang Y,Tang H,Kang X,Yang M,Gustafsson JÅ,Fan X,Tang K
Published figure using Estrogen Receptor beta polyclonal antibody (Product # PA1-313) in Immunohistochemistry
Fri Nov 01 00:00:00 EDT 2019
Effect of estradiol on fibroblasts from postmenopausal idiopathic carpal tunnel syndrome patients.
Journal of cellular physiology
Yamanaka Y,Menuki K,Tajima T,Okada Y,Kosugi K,Zenke Y,Sakai A
Published figure using Estrogen Receptor beta polyclonal antibody (Product # PA1-313) in Immunohistochemistry
Thu Nov 01 00:00:00 EDT 2018
Effect of estradiol on fibroblasts from postmenopausal idiopathic carpal tunnel syndrome patients.
Journal of cellular physiology
Yamanaka Y,Menuki K,Tajima T,Okada Y,Kosugi K,Zenke Y,Sakai A
Published figure using Estrogen Receptor beta polyclonal antibody (Product # PA1-313) in Immunohistochemistry
Thu Nov 01 00:00:00 EDT 2018
Targeting Estrogen Receptor Beta in a Phase 2 Study of High-Dose Estradiol in Metastatic Triple-Negative Breast Cancer: A Wisconsin Oncology Network Study.
Clinical breast cancer
Wisinski KB,Xu W,Tevaarwerk AJ,Saha S,Kim K,Traynor A,Dietrich L,Hegeman R,Patel D,Blank J,Harter J,Burkard ME
PA1-313 was used in immunohistochemistry - paraffin section to analyze a Wisconsin oncology network study of metastatic triple-negative breast cancer where targeting estrogen receptor beta in a phase 2 study of high-dose estradiol
Mon Aug 01 00:00:00 EDT 2016
Targeting Estrogen Receptor Beta in a Phase 2 Study of High-Dose Estradiol in Metastatic Triple-Negative Breast Cancer: A Wisconsin Oncology Network Study.
Clinical breast cancer
Wisinski KB,Xu W,Tevaarwerk AJ,Saha S,Kim K,Traynor A,Dietrich L,Hegeman R,Patel D,Blank J,Harter J,Burkard ME
PA1-313 was used in immunohistochemistry - paraffin section to analyze a Wisconsin oncology network study of metastatic triple-negative breast cancer where targeting estrogen receptor beta in a phase 2 study of high-dose estradiol
Mon Aug 01 00:00:00 EDT 2016
Prognostic significance of full-length estrogen receptor beta expression in stage I-III triple negative breast cancer.
American journal of translational research
Shanle EK,Onitilo AA,Huang W,Kim K,Zang C,Engel JM,Xu W,Wisinski KB
PA1-313 was used in immunohistochemistry and western blot to examine the association between ERbeta1 and Ki67 expression in non-metastatic triple negative breast cancer
Wed Sep 02 00:00:00 EDT 2015
Prognostic significance of full-length estrogen receptor beta expression in stage I-III triple negative breast cancer.
American journal of translational research
Shanle EK,Onitilo AA,Huang W,Kim K,Zang C,Engel JM,Xu W,Wisinski KB
PA1-313 was used in immunohistochemistry and western blot to examine the association between ERbeta1 and Ki67 expression in non-metastatic triple negative breast cancer
Wed Sep 02 00:00:00 EDT 2015
Histone H3 Lysine 27 demethylases Jmjd3 and Utx are required for T-cell differentiation.
Nature communications
Manna S,Kim JK,Baugé C,Cam M,Zhao Y,Shetty J,Vacchio MS,Castro E,Tran B,Tessarollo L,Bosselut R
PA1-313 was used in immunohistochemistry and western blot to test if H3K27Me3 demethylation affects transcription and cell differentiation
Wed Sep 02 00:00:00 EDT 2015
Characterization of estrogen response element binding proteins as biomarkers of breast cancer behavior.
Clinical biochemistry
Kruer TL,Cummins TD,Powell DW,Wittliff JL
PA1-313 was used in EMSA to study the prognostic value of estrogen resonse element binding proteins in breast cancer
Fri Nov 01 00:00:00 EDT 2013
Morphologic transformation of human breast epithelial cells MCF-10A: dependence on an oxidative microenvironment and estrogen/epidermal growth factor receptors.
Cancer cell international
Yusuf R,Frenkel K
Published figure using Estrogen Receptor beta polyclonal antibody (Product # PA1-313) in Western Blot
Wed Sep 01 00:00:00 EDT 2010
Prepubertal physical activity up-regulates estrogen receptor beta, BRCA1 and p53 mRNA expression in the rat mammary gland.
Breast cancer research and treatment
Wang M,Yu B,Westerlind K,Strange R,Khan G,Patil D,Boeneman K,Hilakivi-Clarke L
PA1-313 was used in immunohistochemistry to study the effects of physical activity on the expression of the tumor suppressor genes BRCA1, p53 and estrogen receptor-beta in prepupertal rats and the significance for breast cancer
Fri May 01 00:00:00 EDT 2009
Estrogen increases survival in an orthotopic model of glioblastoma.
Journal of neuro-oncology
Barone TA,Gorski JW,Greenberg SJ,Plunkett RJ
PA1-313 was used in western blot to investigate the effect of estrogen on glioblastoma
Thu Oct 01 00:00:00 EDT 2009
Changes in biomarkers of estrogen receptor and growth factor signaling pathways in MCF-7 tumors after short- and long-term treatment with soy and flaxseed.
The Journal of steroid biochemistry and molecular biology
Power KA,Chen JM,Saarinen NM,Thompson LU
PA1-313 was used in immunohistochemistry to study the effects of soy and flaxseed on tumor growth and estrogen receptor and growth factor signaling pathways in human breast cancer cells using a murine xenograft model
Sat Nov 01 00:00:00 EDT 2008
Differing transcriptional responses to pulsed or continuous estradiol exposure in human umbilical vein endothelial cells.
The Journal of steroid biochemistry and molecular biology
Li J,Wang H,Johnson SM,Horner-Glister E,Thompson J,White IN,Al-Azzawi F
PA1-313 was used in western blot to study the effect of estradiol on transcriptional responses in human umbilical vein endothelial cells
Tue Jul 01 00:00:00 EDT 2008
Changes in transcription profile and cytoskeleton morphology in pelvic ligament fibroblasts in response to stretch: the effects of estradiol and levormeloxifene.
Molecular human reproduction
Ewies AA,Elshafie M,Li J,Stanley A,Thompson J,Styles J,White I,Al-Azzawi F
PA1-313 was used in western blot to investigate the effect of mechanical stretch on gene transcription and cytoskeletonal organization in pelvic ligament fibroblasts
Fri Feb 01 00:00:00 EST 2008
Estrogen receptor beta displays cell cycle-dependent expression and regulates the G1 phase through a non-genomic mechanism in prostate carcinoma cells.
Cellular oncology : the official journal of the International Society for Cellular Oncology
Hurtado A,Pinós T,Barbosa-Desongles A,López-Avilés S,Barquinero J,Petriz J,Santamaria-Martínez A,Morote J,de Torres I,Bellmunt J,Reventós J,Munell F
PA1-313 was used in Western Blotting to demonstrate that, in LNCaP prostate cancer cells, both ERbeta isoforms are differentially expressed during the cell cycle and that ERbeta regulates the G1 phase by a non-genomic mechanism.
Fri Oct 10 00:00:00 EDT 2008
Nuclear estrogen receptor beta in lung cancer: expression and survival differences by sex.
Clinical cancer research : an official journal of the American Association for Cancer Research
Schwartz AG,Prysak GM,Murphy V,Lonardo F,Pass H,Schwartz J,Brooks S
PA1-313 was used in immunohistochemistry to demonstrate the sex-related expression of nuclear estrogen receptor beta in lung cancer.
Sat Oct 15 00:00:00 EDT 2005
Estrogen receptors in the uterus and ovarian follicles of gilts treated with dihydrotestosterone.
Domestic animal endocrinology
Cárdenas H,Pope WF
PA1-313 was used in immunohistochemistry to investigate the expression of estrogen receptor alpha and beta in the uterus and ovarian follicles
Sat Oct 01 00:00:00 EDT 2005
Binding of estrogen receptor beta to estrogen response element in situ is independent of estradiol and impaired by its amino terminus.
Molecular endocrinology (Baltimore, Md.)
Huang J,Li X,Maguire CA,Hilf R,Bambara RA,Muyan M
PA1-313 was used in immunohistochemistry to study the contribution of distinct amino termini of estrogen receptors towards their DNA response element binding and subsequent transcription efficacies.
Tue Nov 01 00:00:00 EST 2005
Oestrogen receptor beta: what it means for patients with breast cancer.
The Lancet. Oncology
Speirs V,Carder PJ,Lane S,Dodwell D,Lansdown MR,Hanby AM
PA1-313 was used in immunohistochemistry to review the significance of oestrogen receptor-beta in breast cancer patients
Mon Mar 01 00:00:00 EST 2004
Single-chain estrogen receptors (ERs) reveal that the ERalpha/beta heterodimer emulates functions of the ERalpha dimer in genomic estrogen signaling pathways.
Molecular and cellular biology
Li X,Huang J,Yi P,Bambara RA,Hilf R,Muyan M
PA1-313 was used in western blot to study the function of estrogen receptor alpha and beta heterodimer.
Wed Sep 01 00:00:00 EDT 2004
Single-chain estrogen receptors (ERs) reveal that the ERalpha/beta heterodimer emulates functions of the ERalpha dimer in genomic estrogen signaling pathways.
Molecular and cellular biology
Li X,Huang J,Yi P,Bambara RA,Hilf R,Muyan M
PA1-313 was used in western blot to study the function of estrogen receptor alpha and beta heterodimer.
Wed Sep 01 00:00:00 EDT 2004
Single-chain estrogen receptors (ERs) reveal that the ERalpha/beta heterodimer emulates functions of the ERalpha dimer in genomic estrogen signaling pathways.
Molecular and cellular biology
Li X,Huang J,Yi P,Bambara RA,Hilf R,Muyan M
PA1-313 was used in western blot to study the function of estrogen receptor alpha and beta heterodimer.
Wed Sep 01 00:00:00 EDT 2004
Single-chain estrogen receptors (ERs) reveal that the ERalpha/beta heterodimer emulates functions of the ERalpha dimer in genomic estrogen signaling pathways.
Molecular and cellular biology
Li X,Huang J,Yi P,Bambara RA,Hilf R,Muyan M
PA1-313 was used in western blot to study the function of estrogen receptor alpha and beta heterodimer.
Wed Sep 01 00:00:00 EDT 2004
Estrogen regulates adrenal angiotensin AT1 receptors by modulating AT1 receptor translation.
Endocrinology
Wu Z,Maric C,Roesch DM,Zheng W,Verbalis JG,Sandberg K
PA1-313 was used in immunohistochemistry to investigate the molecular mechanisms for estrgen regulation of AT1R gene expression in the rat adrenal gland.
Tue Jul 01 00:00:00 EDT 2003
17Beta-estradiol up-regulates vascular endothelial growth factor receptor-2 expression in human myometrial microvascular endothelial cells: role of estrogen receptor-alpha and -beta.
The Journal of clinical endocrinology and metabolism
Gargett CE,Zaitseva M,Bucak K,Chu S,Fuller PJ,Rogers PA
PA1-313 was used in western blot to investigate the role of estrogen receptor-alpha and -beta in human myometrial microvascular endothelial cells.
Sun Sep 01 00:00:00 EDT 2002
17-Beta-estradiol regulates expression of genes that function in macrophage activation and cholesterol homeostasis.
The Journal of steroid biochemistry and molecular biology
Kramer PR,Wray S
PA1-313 was used in western blot to investigate the regulation of 17-beta-estradiol on the expression of genes functioning in macrophage activation and cholesterol metabolism
Mon Jul 01 00:00:00 EDT 2002
Loss of estrogen receptor beta expression in malignant human prostate cells in primary cultures and in prostate cancer tissues.
The Journal of clinical endocrinology and metabolism
Pasquali D,Rossi V,Esposito D,Abbondanza C,Puca GA,Bellastella A,Sinisi AA
PA1-313 was used in western blot to investigate the expression of ER beta and alpha in malignant human prostate cells
Tue May 01 00:00:00 EDT 2001
Different regulation of oestrogen receptors alpha and beta in the human cervix at term pregnancy.
Molecular human reproduction
Wang H,Stjernholm Y,Ekman G,Eriksson H,Sahlin L
PA1-313 was used in immunohistochemistry to study the expression change of the two ERs in the cervix from women at term pregnancy and immediately post-partum and to try to find its relationship with cervical ripening
Thu Mar 01 00:00:00 EST 2001
Estrogen receptors in the normal adult and developing human inner ear and in Turner's syndrome.
Hearing research
Stenberg AE,Wang H,Fish J,Schrott-Fischer A,Sahlin L,Hultcrantz M
PA1-313 was used in immunohistochemistry to investigate the expression and activity of estrogen receptors in human sensory tissues
Sun Jul 01 00:00:00 EDT 2001
Estrogen receptor beta is coexpressed with ERalpha and PR and associated with nodal status, grade, and proliferation rate in breast cancer.
The American journal of pathology
Järvinen TA,Pelto-Huikko M,Holli K,Isola J
PA1-313 was used in immunohistochemistry the study the expression of estrogen receptor beta in normal and pathological human tissues.
Sat Jan 01 00:00:00 EST 2000
Estrogen receptor beta is coexpressed with ERalpha and PR and associated with nodal status, grade, and proliferation rate in breast cancer.
The American journal of pathology
Järvinen TA,Pelto-Huikko M,Holli K,Isola J
PA1-313 was used in immunohistochemistry the study the expression of estrogen receptor beta in normal and pathological human tissues.
Sat Jan 01 00:00:00 EST 2000
Identification and developmental expression of the estrogen receptor alpha and beta in the baboon fetal adrenal gland.
Endocrinology
Albrecht ED,Babischkin JS,Davies WA,Leavitt MG,Pepe GJ
PA1-313 was used in immunohistochemistry to study the expression of the estrogen receptor alpha and beta in the adrenal gland
Wed Dec 01 00:00:00 EST 1999