Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: LHYSDPSPS AWGNLYGAYV PSLALSTLNS CVDPFIYYYV SAEFRDKVRA GLFQRSPGDT VASKASAEGG SRGMGTHSSL LQ (305-385 aa encoded by BC074782)
Coagulation factor II (thrombin) receptor-like 3 (F2RL3) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL3 is also a member of the protease-activated receptor family. F2RL3 is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. F2RL3 is activated by thrombin and trypsin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coagulation factor II (thrombin) receptor-like 3; Coagulation factor II receptor-like 3; PAR-4; protease-activated receptor-4; Proteinase-activated receptor 4; Thrombin receptor-like 3
Gene Aliases: F2RL3; PAR4
UniProt ID: (Human) Q96RI0, (Mouse) O88634
Entrez Gene ID: (Human) 9002, (Mouse) 14065
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support