Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SCKDQLQSY ICFCLPAFEG RNCETHKDDQ LICVNENGGC EQYCSDHTGT KRSCRCHEGY SLLADGVSCT PTVEYPCGKI PILEKRNASK PQGRIVGGKV CPKGECPWQV LLLVNGAQLC GGTLINTIWV VSAAHCFDKI KNWRNLIAVL GEHDLSEHDG DEQSRRVAQV IIPSTYVPGT TNHDIALLRL HQPVVLTDHV VPLCLPERTF SERTLAFVRF SLVSGWGQLL DRGATALELM VLNVPRLMTQ DCLQQSRKVG DSPNITEYMF CAGYSDGSKD SCKGDSGGPH ATHYRGTWYL TGIVSWGQGC ATVGHFGVYT RVSQYIEWLQ KLMRSEPRPG VLLRAPFP (120-466 aa encoded by BC130468)
Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; Factor 7; Factor7; FVII coagulation protein; Proconvertin; Serum prothrombin conversion accelerator; SPCA
Gene Aliases: AI132620; Cf7; F7; FVII; SPCA
UniProt ID: (Mouse) P70375, (Human) P08709
Entrez Gene ID: (Mouse) 14068, (Human) 2155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support