Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAPAPPPAA SFSPSEVQRR LAAGACWVRR GARLYDLSSF VRHHPGGEQL LRARAGQDIS ADLDGPPHRH SANARRWLEQ YYVGELRGEQ QGSMENEPVA LEETQKTDPA MEPRFKVVDW DKDLVDWRKP LLWQVGHLGE KYDEWVHQPV TRPIRLFHSD LIEGL (1-164 aa encoded by BC002679)
FA2H is a protein that catalyzes the synthesis of 2-hydroxysphingolipids, a subset of sphingolipids that contain 2-hydroxy fatty acids. Sphingolipids play roles in many cellular processes and their structural diversity arises from modification of the hydrophobic ceramide moiety, such as by 2-hydroxylation of the N-acyl chain, and the existence of many different head groups.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fatty acid 2-hydroxylase; Fatty acid alpha-hydroxylase; fatty acid hydroxylase domain containing 1; Fatty acid hydroxylase domain-containing protein 1; FLJ25287; spastic paraplegia 35 (autosomal recessive); WD repeat domain 59
Gene Aliases: FA2H; FAAH; FAH1; FAXDC1; G630055L08Rik; RGD1310347; SCS7; SPG35; Wdr59
UniProt ID: (Human) Q9H1A5, (Rat) Q2LAM0, (Mouse) Q5MPP0
Entrez Gene ID: (Human) 79152, (Rat) 307855, (Mouse) 338521
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support