Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: YLIMLPFRF TYYTILDIFR FALRFIRPDP RSRVTDPVGD IVSFMHSFEE KYGRAHPVFY QGTYSQALND AKRELRFLLV YLHGDDHQDS DEFCRNTLCA PEVISLINTR MLFWACSTNK PEGYRVSQAL RENTYPFLAM IMLKDRRMTV VGRLEGLIQP DDLINQLTFI MDANQTYLVS ERLEREERNQ TQVLRQQQDE AYLASLRADQ EKERKKREER ERKRRKEEEV QQQKLAEERR RQNLQEEKER KLECLPPEPS PDDPESVKII FKLPNDSRVE RRFHFSQSLT VIHDFLFSLK ESPEKFQIEA NFPRRVLPCI PSEEWPNPPT LQEAGLSHTE VLFVQDLTDE (97-445 aa encoded by BC014001)
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: expressed in T-cells and eosinophils in atopic dermatitis; FAS-associated factor 2; UBX domain containing 8; UBX domain protein 3B; UBX domain-containing protein 3B; UBX domain-containing protein 8
Gene Aliases: ETEA; FAF2; KIAA0887; UBXD8; UBXN3B
UniProt ID: (Human) Q96CS3
Entrez Gene ID: (Human) 23197
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support