Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
This target displays homology in the following species: Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%
RFLNB is a protein coding gene involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in the formation of cartilaginous skeletal elements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 101, member B; MGC45871; protein FAM101B; Refilin-B; RefilinB; Regulator of filamin protein B
Gene Aliases: CFM1; RFLNB
UniProt ID: (Human) Q8N5W9
Entrez Gene ID: (Human) 359845
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support