Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
The function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 134, member B; protein FAM134B; Reticulophagy receptor 1; Reticulophagy regulator 1
Gene Aliases: 1810015C04Rik; AU015349; FAM134B; JK-1; JK1; RETREG1
UniProt ID: (Human) Q9H6L5, (Rat) Q5FVM3, (Mouse) Q8VE91
Entrez Gene ID: (Human) 54463, (Rat) 619558, (Mouse) 66270
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support