Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: LGLPPVSPRD CVKDAVAAEV FDHVWTNMVE LLEELIRKHW ETTLTEGKKQ RETLKVAGNR FPHVLVPHAH ADGASGPPSG HAEAHGISLA SRLNPPQIHH FSSSF
Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 58%.
FAM149A is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP564J102; family with sequence similarity 149, member A; Protein FAM149A
Gene Aliases: FAM149A; MST119; MSTP119
UniProt ID: (Human) A5PLN7
Entrez Gene ID: (Human) 25854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support