Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FAM151B is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AASA9217; f151; family with sequence similarity 151, member B; Protein FAM151B
Gene Aliases: 4930405M20Rik; FAM151B; UNQ9217; UNQ9217/PRO28685
UniProt ID: (Human) Q6UXP7, (Mouse) D3YUE4
Entrez Gene ID: (Human) 167555, (Mouse) 73942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support