Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: DEVHQNQILR ELYLKELRTQ KLHTQYHVNP
FAM183A is a protein coding gene. Gene ontology (GO) annotation include ciliary base.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 144; family with sequence similarity 183, member A; hCG23177; LOC440585; Protein FAM183A
Gene Aliases: CFAP144; FAM183A
UniProt ID: (Human) A6NL82
Entrez Gene ID: (Human) 440585
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support