Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Human: 100%
May negatively regulate STAT3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 220, member A; Protein FAM220A; STAT3-interacting protein as a repressor
Gene Aliases: ACPIN1; C7orf70; FAM220A; SIPAR
UniProt ID: (Human) Q7Z4H9
Entrez Gene ID: (Human) 84792
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support