Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Cytoplasmic non-canonical poly(A) RNA polymerase that catalyzes the transfer of one adenosine molecule from an ATP to an mRNA poly(A) tail bearing a 3'-OH terminal group and participates in the cytoplasmic polyadenylation. Polyadenylates mRNA encoding extracellular matrix constituents and other genes crucial for bone mineralization and during osteoblast mineralization, mainly focuses on ER-targeted mRNAs (By similarity).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 46, member A; HBV X-transactivated gene 11 protein; HBV XAg-transactivated protein 11; retinal expressed gene C6orf37; Terminal nucleotidyltransferase 5A
Gene Aliases: C6orf37; FAM46A; TENT5A; XTP11
UniProt ID: (Human) Q96IP4
Entrez Gene ID: (Human) 55603
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support