Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This antibody detects all the members of FAM72.
Immunogen sequence: MSTNICSFK DRCVSILCCK FCKQVLSSRG MKAVLLADTE IDLFSTDIPP TNAVDFTGRC YFTKICKCKL KDIACLKCGN IVGYHVIVPC SSCLLSCNNG HFWMFHSQAV YDINRLDSTG VNVLLWGNLP EIEESTDEDV LNISAEECIR (1-149 aa encoded by BC035696)
FAM72A, also named as LMIP and UGENE, is a mitochondrial protein which regulates cellular reactive oxygen species metabolism. It is up-regulated in malignant colon cancers, compared to normal colon and colon adenomas. FAM72 has some members A/B/C/D with higher homology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 72, member A; Latent membrane protein 1-induced protein; LMP1-induced protein; MGC57827; Protein FAM72A; RP11-312O7.1
Gene Aliases: 2700049P18Rik; FAM72A; LMPIP; P17; UGENE
UniProt ID: (Human) Q5TYM5, (Mouse) Q8BFZ8
Entrez Gene ID: (Human) 729533, (Mouse) 108900
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support