Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with X. laevis and X. tropicalis
FAM76B is an uncharacterized protein belonging to the FAM76 family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 76, member B; Protein FAM76B
Gene Aliases: 2810485I05Rik; C78303; FAM76B
UniProt ID: (Human) Q5HYJ3, (Mouse) Q80XP8
Entrez Gene ID: (Human) 143684, (Mouse) 72826
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support