Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene plays an important role in the structural development and calcification of tooth enamel. Defects in this gene are a cause of amelogenesis imperfecta type 3 (AI3).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAM83H variant 1; FAM83H variant 2; family with sequence similarity 83, member H; FLJ46072; Protein FAM83H
Gene Aliases: AA409316; AI3; FAM83H
UniProt ID: (Human) Q6ZRV2, (Mouse) Q148V8
Entrez Gene ID: (Human) 286077, (Mouse) 105732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support