Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MSRQTTSVGS SCLDLWREKN DRLVRQAKVA QNSGLTLRRQ QLAQDALEGL RGLLHSLQGL PAAVPVLPLE LTVTCNFIIL RASLAQGFTE DQAQDIQRSL ERVLETQEQQ GPRLEQGLRE LWDSVLRASC LLPELLSALH RLVGLQAALW LSADRLGDLA LLLETLNGSQ SGASKDLLLL LKTWSPPAEE LDAPLTLQDA QGLKDVLLTA FAYRQGLQEL ITGNPDKALS SLHEAASGLC PRPVLVQVYT ALGSCHRKMG NPQRALLYLV AALKEGSAWG PPLLEASRLY QQLGDTTAEL ESLELLVEAL NVPCSSKAPQ FLIEVELLLP PPDLASPLHC GTQSQTKHIL ASRCLQTGRA GDAAEHYLDL LALLLDSSEP RFSPPPSPPG PCMPEVFLEA AVALIQAGRA QDALTLCEEL LSRTSSLLPK MSRLWEDARK GTKELPYCPL WVSATHLLQG QAWVQLGAQK VAISEFSRCL ELLFRATPEE KEQGAAFNCE QGCKSDAALQ QLRAAALISR GLEWVASGQD TKALQDFLLS VQMCPGNRDT YFHLLQTLKR LDRRDEATAL WWRLEAQTKG SHEDALWSLP LYLESYLSWI RPSDRDAFLE EFRTSLPKSC DL
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. FANCG is the protein for complementation group G.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA repair protein XRCC9; Fanconi anemia group G protein; Fanconi anemia, complementation group G; Protein FACG; truncated Fanconi anemia group G protein; X-ray repair complementing defective repair in Chinese hamster cells 9; X-ray repair, complementing defective, in Chinese hamster, 9
Gene Aliases: FAG; FANCG; XRCC9
UniProt ID: (Human) O15287
Entrez Gene ID: (Human) 2189
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support