Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Control: Rat colon tissue lysate, 293T cell lysate, SH-SY5Y, rat testis tissue, human small intestine tissue, mouse cerebellum tissue, SiHa.
Subcellular Location: Mitochondrion matrix, Mitochondrion.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ236A3.1 (phenylalanine-tRNA synthetase); dJ520B18.2; dJ520B18.2 (FARS1 (phenylalanine-tRNA synthetase)); mitochondrial PHERS; pheny; phenylalanine translase; phenylalanine tRNA ligase 2, mitochondrial; Phenylalanine--tRNA ligase, mitochondrial; phenylalanine-tRNA synthetase 1 (mitochondrial); phenylalanine-tRNA synthetase 2 (mitochondrial); Phenylalanyl-tRNA synthetase; phenylalanyl-tRNA synthetase, mitochondrial; PheRS
Gene Aliases: COXPD14; FARS1; FARS2; HSPC320; PheRS
UniProt ID: (Human) O95363, (Rat) Q6AYQ3, (Mouse) Q99M01
Entrez Gene ID: (Human) 10667, (Rat) 306879, (Mouse) 69955
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support