Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MADGQVAELL LRRLEASDGG LDSAELAAEL GMEHQAVVGA VKSLQALGEV IEAELRSTKH WELTAEGEEI AREGSHEARV FRSIPPEGLA QSELMRLPSG KVGFSKAMSN KWIRVDKSAA DGPRVFRVVD SMEDEVQRRL QLVRGGQAEK LGEKERSELR KRKLLAEVTL KTYWVSKGSA FSTSISKQET ELSPEMISSG SWRDRPFKPY NFLAHGVLPD SGHLHPLLKV RSQFRQIFLE MGFTEMPTDN FIESSFWNFD ALFQPQQHPA RDQHDTFFLR DPAEALQLPM DYVQRVKRTH SQGGYGSQGY KYNWKLDEAR KNLLRTHTTS ASARALYRLA QKKPFTPVKY FSIDRVFRNE TLDATHLAEF HQIEGVVADH GLTLGHLMGV LREFFTKLGI TQLRFKPAYN PYTEPSMEVF SYHQGLKKWV EVGNSGVFRP EMLLPMGLPE NVSVIAWGLS LERPTMIKYG INNIRELVGH KVNLQMVYDS PLCRLDAEPR PPPTQEAA
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CML33; phenylalanine tRNA ligase 1, alpha, cytoplasmic; phenylalanine--tRNA ligase alpha chain; Phenylalanine--tRNA ligase alpha subunit; phenylalanine-tRNA synthetase alpha-subunit; phenylalanine-tRNA synthetase-like, alpha subunit; phenylalanyl-tRNA synthetase alpha chain; Phenylalanyl-tRNA synthetase alpha subunit; phenylalanyl-tRNA synthetase, alpha subunit; phenylalanyl-tRNA synthetase-like, alpha subunit; PheRS
Gene Aliases: CML33; FARS; FARSA; FARSL; FARSLA; FRSA; PheHA
UniProt ID: (Human) Q9Y285
Entrez Gene ID: (Human) 2193
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support