Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKPGFSPRGG GFGGRGGFGD RGGRGGRGGF GGGRGRGGGF RGRGRGGGGG GGGGGGGGRG GGGFHSGGNR GRGRGGKRGN QSGKNVMVEP HRHEGVFICR GKEDALVTKN LVPGESVYGE KRVSISEGDD KIEYRAWNPF RSKLAAAILG GVDQIHIKPG AKVLYLGAAS GTTVSHVSDI VGPDGLVYAV EFSHRSGRDL INLAKKRTNI IPVIEDARHP HKYRMLIAMV DVIFADVAQP DQTRIVALNA HTFLRNGGHF VISIKANCID TTASAEAVFA SEVKKMQQEN MKPQEQLTLE PYERDHAVVV GVYRPPPKVK N
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; Histone-glutamine methyltransferase; RNA, U3 small nucleolar interacting protein 1; rRNA 2'-O-methyltransferase fibrillarin; U6 snRNA 2'-O-methyltransferase fibrillarin
Gene Aliases: FBL; FIB; FIB1; FLRN; RNU3IP1
UniProt ID: (Human) P22087
Entrez Gene ID: (Human) 2091
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support