Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein with an N-terminal filamin-binding domain, a central proline-rich domain, and, multiple C-terminal LIM domains. This protein localizes at cell junctions and may link cell adhesion structures to the actin cytoskeleton. This protein may be involved in the assembly and stabilization of actin-filaments and likely plays a role in modulating cell adhesion, cell morphology and cell motility. This protein also localizes to the nucleus and may affect cardiomyocyte differentiation after binding with the CSX/NKX2-5 transcription factor. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CSX-associated LIM; FBLP-1; Filamin-binding LIM protein 1; filamin-binding LIM protein-1; MIG2-interacting protein; Migfilin; mitogen-inducible 2 interacting protein; Mitogen-inducible 2-interacting protein
Gene Aliases: CAL; FBLIM1; FBLP-1; FBLP1
UniProt ID: (Human) Q8WUP2
Entrez Gene ID: (Human) 54751
Molecular Function: kinase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support