Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: FLECQNSPAR ITHYQLNFQT GLLVPAHIFR IGPAPAFTGD TIALNIIKGN EEGYFGTRRL NAYTGVVYLQ RAVLEPRDFA LDVEMKLWRQ GSVTTFLAKM HIFFTTFAL
This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FIBL-2; FIBL2; Fibulin-2; Fibulin2
Gene Aliases: FBLN2
UniProt ID: (Human) P98095
Entrez Gene ID: (Human) 2199
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support