Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MKRGGRDSDR NSSEEGTAEK SKKLRTTNEH SQTCDWGNLL QDIILQVFKY LPLLDRAHAS QVCRNWNQVF HMPDLWRCFE FELNQPATSY LKATHPELIK
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats and is localized in the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box and leucine-rich repeat protein 3; F-box and leucine-rich repeat protein 3A; F-box protein Fbl3a; F-box/LRR-repeat protein 3; F-box/LRR-repeat protein 3A; Protein after-hours; Protein overtime
Gene Aliases: Afh; AU041772; AW212966; FBK; FBL3; FBL3A; FBXL3; FBXL3A; Ovtm; Play68
UniProt ID: (Human) Q9UKT7, (Mouse) Q8C4V4
Entrez Gene ID: (Human) 26224, (Mouse) 50789, (Rat) 306129
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support