Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: AEDKELEAK IAVVEKYNIR IPELVQRIEK CHIEDLDFAE YILGTVHKAK GLEFDTVHVL DDFVKVPCAR HNLPQLPHFR VESFSEDEWN LLYVAVTRAK KRLIMTKSLE NILTLAGEYF LQAELTSNVL KTGVVRCCVG QCNNAIPVDT VLTMKKLPIT YSNRKENKGG YLCHSCAEQR IGPLAFLTAS PEQVRAMERT VENIVLPRHE ALLFLVF (693-1043 aa encoded by BC032674)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA 3'-5' helicase 1; F-box DNA helicase 1; F-box only protein 18; F-box only protein, helicase, 18; hFBH1
Gene Aliases: AU015756; FBH1; FBX18; FBXO18; hFBH1
UniProt ID: (Human) Q8NFZ0, (Mouse) Q8K2I9
Entrez Gene ID: (Human) 84893, (Mouse) 50755, (Rat) 291293
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support