Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: GGRQEEFRTW LREEWGRTLE DIFHEHMQEL ILMKFIYTSQ YDNCLTYRRI YLPPSRPDDL IKPGLFKGTY GSHGLEIVML SFHGRRARGT KITGDPNIPA GQQTVEIDLR HRIQLPDLEN QRNFNELSRI VLEVRERVRQ EQQEGGHEAG EGRGRQGPRE SQPSPAQPRA EAPSKGPDGT PGEDGGEPGD AVAAAEQPAQ CGQGQPFVLP VGVSSRNEDY PRTCRMCFYG TGLIAGHGFT SPERTPGVFI LFDEDRFGFV WLELKSFSLY SRVQATFRNA DAPSPQAFDE MLKNIQSLTS; Positive Samples: Mouse brain, Rat brain
Members of the F-box protein family, such as FBXO31, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp434B027; DKFZp434J1815; F-box only protein 14; F-box only protein 31; FLJ22477; MGC15419; MGC9527
Gene Aliases: 1110003O08Rik; 2310046N15Rik; Fbx14; Fbxo14; Fbxo31; MNCb-2609; RGD1561069
UniProt ID: (Rat) B2RYN2, (Mouse) Q3TQF0
Entrez Gene ID: (Rat) 498959, (Mouse) 76454
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support