Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGKARRSPPG HHRHCEGCFN RHCHIPVEPN TSCLVISCHL LCGATFHMCK EAEHQLLCPL EQVPCLNSEY GCPLSMSRHK LAKHLQVCPA SVVCCSMEWN RWPNVDSETT LHENIMKETP SEECLDTALA LQDQKVLFRS LKMVELFPET REATEEEPTM NGETSVEEMG GAVGGVDIGL VPHGLSATNG EMAELSQEER EVLAKTKEGM DLVKFGQWEN IFSKEHAASA LTNSSASCES KNKNDSEKEQ ISSGHNMVEG EGAPKKKEPQ ENQKQQDVRT AMETTGLAPW QDGVLERLKT AVDAKDYNMY LVHNGRMLIH FGQMPACTPK ERDFVYGKLE AQEVKTVYTF KVPVSYCGKR ARLGDAMLSC KPSEHKAVDT SDLGITVEDL PKSDLIKTTL QCALERELKG HVISESRSID GLFMDFATQT YNFEPEQFSS GTVLADLTAA TPGGLHVELH SECVTRRHNK SSSAFTFTCN KFFRRDEFPL HFKNVHTDIQ SCLNGWFQHR CPLAYLGCTF VQNHFRPPGQ KAKVIYSQEL KTFAIKPEVA PELSEGRKNN HLLGHGGKSQ NSLTSLPLEI LKYIAGFLDS VSLAQLSQVS VLMRNICATL LQERGMVLLQ WKKKRYSHGG TSWRVHREIW QFSSLFSKIK SWEFNEVTSM SEHLKSCPFN IVEHKTDPIL LTSMCQPREQ ARESLVSTFR IRPRGRYVS
Members of the F-box protein family, such as FBXO40, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: burs-beta; F-box only protein 40; Muscle disease-related protein
Gene Aliases: FBX40; FBXO40; KIAA1195
UniProt ID: (Human) Q9UH90
Entrez Gene ID: (Human) 51725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support