Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
In Western blots, approximately 5% cross-reactivity with recombinant human (rh) IRTA5 is observed and less than 2% cross-reactivity with rhITRA1, rhIRTA2, and rhIRTA3 is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
The FCRL2 gene encodes the protein Fc receptor-like 2, which is part of the Fc receptor-like family involved in immune system regulation. FCRL2 is primarily expressed on the surface of B lymphocytes, where it contributes to the modulation of B cell activation and immune responses. As a member of the Fc receptor-like family, FCRL2 is thought to play roles in signaling pathways that influence B cell development, differentiation, and antibody production. It may act as a regulatory receptor that balances immune activation and inhibition, thereby contributing to immune homeostasis and prevention of autoimmunity. The expression of FCRL2 can impact conditions such as autoimmune diseases and B cell-related disorders, where dysregulation of B cell signaling is evident. Research into FCRL2 is ongoing to explore its specific ligand interactions and functional mechanisms, aiming to understand its potential as a therapeutic target for modulating immune responses in diseases characterized by B cell dysfunction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD307b; Fc receptor homolog 2; Fc receptor-like 2; Fc receptor-like protein 2; FcR-like protein 2; FcRH2; IFGP family protein 4; immune receptor translocation-associated protein 4; Immunoglobulin receptor translocation-associated protein 4; immunoglobulin superfamily Fc receptor, gp42; RP11-367J7.6; SH2 domain containing phosphatase anchor protein 1; SH2 domain-containing phosphatase anchor protein 1
Gene Aliases: CD307b; FCRH2; FCRL2; IFGP4; IRTA4; SPAP1; SPAP1A; SPAP1B; SPAP1C; UNQ9236/PRO31998
UniProt ID: (Human) Q96LA5
Entrez Gene ID: (Human) 79368
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support