Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LPQEGTSVL PFWNCDFLSA AFWISLHEDN SNSESLTGGT SQDVEDFLVS FSELSLLKNP GRDGKEEACE GTCGQAKICL RPSLLVHVQD VIEVPDFLSG SLHILSGPVF QKCHILPFTM PAFHETLFIL GVNQNLKDGC LQSLLDHLKG ILDSLLTQTL PESSKLSSLV KFVLQSNGKD YMIRVKTHNF SPDCTEDLII GSVITSATSV IHKDQCFVFV SMNLDLLAML VWCISDWRML WTFDNRFLKN FVPGKIEPFK SHSLYPPCYV HDVSFWIDQK KGFDELEFHT VARAVSQDTI ISIQFLSRFQ HPKTQQVSLC YRLTYQTCDK ALTQQQVASM QSQFRKEIQQ HLYVIPR (269-624 aa encoded by BC127680 )
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FDX-ACDB domain-containing protein 1; Ferredoxin-fold anticodon-binding domain-containing protein 1; hypothetical protein BC006136
Gene Aliases: FDXACB1; hCG_2033039
UniProt ID: (Human) Q9BRP7
Entrez Gene ID: (Human) 91893
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support