Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
FIGURE: 1 / 1
The most common immunoglobulin class is IgG. Each chain is divided into regions or domains consisting of around 110 amino acid residues. The light chain has two domains and the heavy has four. The N-terminal domain at the tip of the arms of the "Y" on both the heavy and light chain are known to be variable in amino acid sequence composition and are thus called variable domains (VL and VH). The other domains are called constant for a similar reason (CL, CH1, CH2, CH3).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AG4; Allergen Cat-1; Allergen FdI; Allergen Fel d I-A; allergen Fel dI; CH1; Major allergen I polypeptide chain 1
Gene Aliases: CH1
UniProt ID: (Cat) P30438
Entrez Gene ID: (Cat) 677877
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support