Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGIQGLAKL IADVAPSAIR ENDIKSYFGR KVAIDASMSI YQFLIAVRQG GDVLQNEEGE TTSHLMGMFY RTIRMMENGI KPVYVFDGKP PQLKSGELAK RSERRAEAEK QLQQAQAAGA EQEVEKFTKR LVKVTKQHND ECKHLLSLMG IPYLDAPSEA EASCAALVKA GKVYAAATED MDCLTFGSPV LMRHLTASEA KKLPIQEFHL SRILQELGLN QEQFVDLCIL LGSDYCESIR GIGPKRAVDL IQKHKSIEEI VRRLDPNKYP VPENWLHKEA HQLFLEPEVL DPESVELKWS EPNEEELIKF MCGEKQFSEE RIRSGVKRLS KSRQGSTQGR LDDFFKVTGS LSSAKRKEPE PKGSTKKKAK TGAAGKFKRG K (1-380 aa encoded by BC000323)
FEN-1 (Flap Endonuclease and Five prime Exonuclease) is a 42 a protein that functions as an endonuclease that cleaves branched DNA structures containing single-stranded 5' flaps or as an exonuclease on DNA nicks and gaps. FEN-1 endonuclease and exonuclease activities are stimulated by an association with proliferating cell nuclear antigen (PCNA) in the nucleus. FEN-1 is believed to have important roles in DNA replication and repair and is required in cell-free DNA replication systems.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNase IV; FEN-1; FEN-1 {ECO:0000255|HAMAP-Rule:MF_03140}; FEN1; fen1 {ECO:0000255|HAMAP-Rule:MF_03140}; Flap endonuclease 1; flap endonuclease 1 {ECO:0000255|HAMAP-Rule:MF_03140}; Flap structure-specific endonuclease 1; flap structure-specific endonuclease 1 {ECO:0000255|HAMAP-Rule:MF_03140}; hFEN-1; Maturation factor 1; maturation factor-1; MF1
Gene Aliases: AW538437; FEN-1; FEN1; MF1; RAD2
UniProt ID: (Human) P39748, (Rat) Q5XIP6
Entrez Gene ID: (Human) 2237, (Mouse) 14156, (Rat) 84490
Molecular Function:
exodeoxyribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support