Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: CPTCGKGFCR NFDLKKHVRK LHDSSLGLAR TPAGEPGTEP PPPLPQQPPM
Sequence homology: Guinea Pig: 79%; Horse: 79%; Human: 100%; Rabbit: 92%; Rat: 100%
Transcription repressor. Involved in the axonal projection and proper termination of olfactory sensory neurons (OSN). Plays a role in rostro-caudal patterning of the diencephalon and in prethalamic formation. Expression is required in OSN to cell-autonomously regulate OSN axon projections. Regulates non-cell-autonomously the layer formation of the olfactory bulb development and the interneurons. May be required for correct rostral migration of the interneuron progenitors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fez family zinc finger protein 1; Zinc finger protein 312B
Gene Aliases: FEZ; FEZF1; HH22; ZNF312B
UniProt ID: (Human) A0PJY2
Entrez Gene ID: (Human) 389549
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support