Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. GPRs translate extracellular signals into intracellular signals (a process called G-protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR41 (G-protein coupled receptor 41), also known as FFAR3 (Free fatty acid receptor 3), is a 346 amino acid multi-pass membrane protein that belongs to the G protein-coupled receptor family. Expressed at high levels in adipose tissue and at lower levels throughout the body, GPR41 functions as a receptor for short chain fatty acids via elevation of intracellular calcium levels and inhibition of adenylyl cyclase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Free fatty acid receptor 3; G protein-coupled receptor 41; G-protein coupled receptor 41; Gpcr41; putative G protein-coupled receptor GPR41
Gene Aliases: FFA3R; FFAR3; Gm478; GPR41; GPR42
UniProt ID: (Human) O14843, (Rat) B2GV46, (Mouse) Q3UFD7
Entrez Gene ID: (Human) 2865, (Rat) 365228, (Mouse) 233080
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support