Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MESGRGSSTP PGPIAALGMP DTGPGSSSLG KLQALPVGPR AHCGDPVSLA AAGDGSPDIG PTGELSGSLK IPNRDSGIDS PSSSVAGENF PCEEGLEAGP SPTVLGAHAE MALDSQVPKV TPQEEADSDV GEEPDSENTP QKADKDAGLA QHSGPQKLLH IAQELLHTEE TYVKRLHLLD QVFCTRLTDA GIPPEVIMGI FSNISSIHRF HGQFLLPELK TRITEEWDTN PRLGDILQKL APFLKMYGEY VKNFDRAVGL VSTWTQRSPL FKDVIHSIQK QEVCGNLTLQ HHMLEPVQRV PRYELLLKDY LKRLPQDAPD RKDAERSLEL ISTAANHSNA AIRKVEKMHK LLEVYEQLGG EEDIVNPANE LIKEGQIQKL SAKNGTPQDR HLFLFNSMIL YCVPKLRLMG QKFSVREKMD ISGLQVQDIV KPNTAHTFII TGRKRSLELQ TRTEEEKKEW IQIIQATIEK HKQNSETFKA FGGAFSQDED PSLSPDMPIT STSPVEPVVT TEGSSGAAGL EPRKLSSKTR RDKEKQSCKS CGETFNSITK RRHHCKLCGA VICGKCSEFK AENSRQSRVC RDCFLTQPVA PESTEKTPTA DPQPSLLCGP LRLSESGETW SEVWAAIPMS DPQVLHLQGG SQDGRLPRTI PLPSCKLSVP DPEERLDSGH VWKLQWAKQS WYLSASSAEL QQQWLETLST AAHGDTAQDS PGALQLQVPM GAAAP
FGD3 gene ontology annotations related to this gene include regulation of Rho protein signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: faciogenital dysplasia 3; FGD1 family, member 3; FLJ00004; FYVE, RhoGEF and PH domain-containing protein 3; Zinc finger FYVE domain-containing protein 5
Gene Aliases: FGD3; ZFYVE5
UniProt ID: (Human) Q5JSP0
Entrez Gene ID: (Human) 89846
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support