Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRALDDMDH EGRDTLAREE LRQGLSELPA IHDLHQGILE ELEERLSNWE SQQKVADVFL AREQGFDHHA THILQFDRYL GLLSENCLHS PRLAAAVREF EQSVQGGSQT AKHRLLRVVQ RLFQYQVLLT DYLNNLCPDS AEYDNTQGAL SLISKVTDRA NDSMEQGENL QKLVHIEHSV RGQGDLLQPG REFLKEGTLM KVTGKNRRPR HLFLMNDVLL YTYPQKDGKY RLKNTLAVAN MKVSRPVMEK VPYALKIETS ESCLMLSASS CAERDEWYGC LSRALPEDYK AQALAAFHHS VEIRERLGVS LGERPPTLVP VTHVMMCMNC GCDFSLTLRR H (923-1262 aa encoded by BC035364)
FGD5 activates CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. It mediates VEGF-induced CDC42 activation. It may regulate proangiogenic action of VEGF in vascular endothelial cells, including network formation, directional movement and proliferation and may play a role in regulating the actin cytoskeleton and cell shape.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FYVE, RhoGEF and PH domain-containing protein 5; Zinc finger FYVE domain-containing protein 23
Gene Aliases: FGD5; ZFYVE23
UniProt ID: (Human) Q6ZNL6
Entrez Gene ID: (Human) 152273
Molecular Function:
G-protein modulator
protein-binding activity modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support