Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NSPQLKSRTG KLRASESPSS LIFYRDGKRK GVPFSRTVSR VESFEDRSRP PFLPLPLTKP RSISFPSADT SDYENIPAMN SDYENIQIPP RRPARAGAFT KLFEDQSRAL STANENDGYV D
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
FGD5 activates CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. It mediates VEGF-induced CDC42 activation. It may regulate proangiogenic action of VEGF in vascular endothelial cells, including network formation, directional movement and proliferation and may play a role in regulating the actin cytoskeleton and cell shape.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FYVE, RhoGEF and PH domain-containing protein 5; Zinc finger FYVE domain-containing protein 23
Gene Aliases: FGD5; ZFYVE23
UniProt ID: (Human) Q6ZNL6
Entrez Gene ID: (Human) 152273
Molecular Function:
G-protein modulator
protein-binding activity modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support