Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: YLGLLSENCL HSPRLAAAVR EFEQSVQGGS QTAKHRLLRV VQRLFQYQVL LTDYLNNLCP DSAEYDNTQG ALSLISKVTD RANDSMEQGE NLQKLVHIEH SVRGQGDLLQ PGREFLKEGT LMKVTGKNRR PRHLFLMNDV LLYTYPQKDG KYRLKNTLAV ANMKVSRPVM EKVPYALKIE TSESCLMLSA SSCAERDEWY G; Positive Samples: Mouse brain; Cellular Location: Cell projection, Cytoplasm, Early endosome, Endoplasmic reticulum, Golgi apparatus, cytoskeleton, ruffle membrane
FGD5 activates CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. It mediates VEGF-induced CDC42 activation. It may regulate proangiogenic action of VEGF in vascular endothelial cells, including network formation, directional movement and proliferation and may play a role in regulating the actin cytoskeleton and cell shape.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FYVE, RhoGEF and PH domain-containing protein 5
Gene Aliases: C330025N11Rik; Fgd5; mFLJ00274; ZFYVE23
UniProt ID: (Mouse) Q80UZ0
Entrez Gene ID: (Mouse) 232237
Molecular Function:
G-protein modulator
protein-binding activity modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support