Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LGQDMVSPEA TNSSSSSFSS PSSAGRHVRS YNHLQGDVRW RKLFSFTKYF LKIEKNGKVS GTKKENCPYS ILEITSVEIG VVAVKAINSN YYLAMNKKGK LYGSKEFNND CKLKERIEEN GYNTYASFNW QHNGRQMYVA LNGKGAPRRG QKTRRKNTSA HFLPMVVHS; Positive Samples: A-549
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Keratinocyte growth factor 2
Gene Aliases: AEY17; BB213776; Fgf-10; Fgf10; Gsfaey17
UniProt ID: (Mouse) O35565
Entrez Gene ID: (Mouse) 14165
Molecular Function:
growth factor
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support