Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Probably involved in nervous system development and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-11; FHF-3; Fibroblast growth factor; Fibroblast growth factor 11; Fibroblast growth factor homologous factor 3
Gene Aliases: FGF-11; FGF11; FHF-3; FHF3
UniProt ID: (Human) Q92914, (Mouse) P70378
Entrez Gene ID: (Human) 2256, (Mouse) 14166
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support